Learn More
MSC Mag-Mate Magnetic Grate Rod Separator, 14 in Long, 1 in Diameter, Ceramic Magnet
Catalog No. 5000513882
Encompass
This magnetic tube separator is designed for custom installations requiring magnetic separation in limited spaces. It features tapped mounting holes at each end for easy bolt-in installation and is suitable for gravity-feed product applications across various industries.
  • Standard lengths available from 4 to 36 inches in 2 inch increments
  • Constructed with 1 inch diameter stainless steel tubes
  • Includes rolled ends with stainless steel plugs
  • Equipped with 1/4-20 tapped end plug mounting holes
  • Options for welded ends and custom lengths
  • Special coatings are available for abrasive products
  • Utilized in industries including food, feed, grain, pharmaceutical, petrochem, and plastics
Catalog No. 50-005-13882 Supplier MSC Supplier No. 64830318
This magnetic tube separator is designed for custom installations requiring magnetic separation in limited spaces. It features tapped mounting holes at each end for easy bolt-in installation and is suitable for gravity-feed product applications across various industries.
  • Standard lengths available from 4 to 36 inches in 2 inch increments
  • Constructed with 1 inch diameter stainless steel tubes
  • Includes rolled ends with stainless steel plugs
  • Equipped with 1/4-20 tapped end plug mounting holes
  • Options for welded ends and custom lengths
  • Special coatings are available for abrasive products
  • Utilized in industries including food, feed, grain, pharmaceutical, petrochem, and plastics