Learn More
MSC Mag-Mate Magnetic Grate Rod Separator, 20 in x 1 in, Ceramic Magnet
Catalog No. 5000512051
Encompass
This magnetic grate rod separator is designed to efficiently remove ferrous contaminants from dry, free-flowing materials. Featuring a durable 304 stainless steel rod and a powerful ceramic magnet, it measures 20 inches in length and 1 inch in diameter, making it suitable for various industrial applications.
  • Features a powerful ceramic magnet
  • Constructed with durable 304 stainless steel rod
  • Measures 20 in long x 1 in diameter
  • Designed for removing ferrous contaminants
  • Suitable for dry, free-flowing materials
  • Ensures product purity and protects machinery
Catalog No. 50-005-12051 Supplier MSC Supplier No. 64830342
This magnetic grate rod separator is designed to efficiently remove ferrous contaminants from dry, free-flowing materials. Featuring a durable 304 stainless steel rod and a powerful ceramic magnet, it measures 20 inches in length and 1 inch in diameter, making it suitable for various industrial applications.
  • Features a powerful ceramic magnet
  • Constructed with durable 304 stainless steel rod
  • Measures 20 in long x 1 in diameter
  • Designed for removing ferrous contaminants
  • Suitable for dry, free-flowing materials
  • Ensures product purity and protects machinery