Learn More
MSC Mag-Mate Magnetic Lifter, 1,000 lb Capacity, Permanent, Stainless Steel
Catalog No. 5000535353
Encompass
A powerful and economical ceramic lift magnet solution designed for thick, non-flexing steel sheets and parts. Features a lightweight and durable stainless steel design. The Creative Lift has a Non-Marring Roller Cam Release to release the magnet from the steel surface. Includes a tall lift lug for easy use with crane hooks and slings.
  • Less torque required to release load
  • Spring return handle
  • "Jack screw" secondary release
  • 3:1 Design factor
  • Non-marring roller cam release
  • Tall lift lug for easy use with crane hooks and slings
  • Lightweight and durable stainless steel design
Catalog No. 50-005-35353 Supplier MSC Supplier No. 06586382
A powerful and economical ceramic lift magnet solution designed for thick, non-flexing steel sheets and parts. Features a lightweight and durable stainless steel design. The Creative Lift has a Non-Marring Roller Cam Release to release the magnet from the steel surface. Includes a tall lift lug for easy use with crane hooks and slings.
  • Less torque required to release load
  • Spring return handle
  • "Jack screw" secondary release
  • 3:1 Design factor
  • Non-marring roller cam release
  • Tall lift lug for easy use with crane hooks and slings
  • Lightweight and durable stainless steel design