Learn More
MSC MAG-MATE Magnetic Sheet Separator, 12-22 ga, 15.25 in Height, Steel
Catalog No. 5000529382
Encompass
The Industrial Magnetics, Inc. MGF15 Magnetic Sheet Separator uses a powerful magnetic field to efficiently separate steel sheet stock, eliminating the need for manual prying and preventing double blanking. This permanent sheet fanner is ideal for applications in automotive, fabrication, welding, appliance, stamping, furniture, and equipment manufacturing.
  • Features a flexible mounting design for versatile use
  • Includes pre-tapped holes for easy mounting
  • Comes with a bolt-on angle base plate and handle with mounting hardware
  • Durable welded construction ensures longevity
  • Equipped with wear strips on the magnetic face
  • Optimized for separating 12 to 22 gauge metal sheets
Catalog No. 50-005-29382 Supplier MSC Supplier No. 06586069
The Industrial Magnetics, Inc. MGF15 Magnetic Sheet Separator uses a powerful magnetic field to efficiently separate steel sheet stock, eliminating the need for manual prying and preventing double blanking. This permanent sheet fanner is ideal for applications in automotive, fabrication, welding, appliance, stamping, furniture, and equipment manufacturing.
  • Features a flexible mounting design for versatile use
  • Includes pre-tapped holes for easy mounting
  • Comes with a bolt-on angle base plate and handle with mounting hardware
  • Durable welded construction ensures longevity
  • Equipped with wear strips on the magnetic face
  • Optimized for separating 12 to 22 gauge metal sheets