Learn More
MSC Mag-Mate Neodymium Disc Magnet, Countersunk, 0.750 in Diameter, N42 Grade
Catalog No. 5000515769
Encompass
This Neodymium disc magnet is designed for strong holding applications. It features a countersunk design for flush mounting and offers a high pull force, making it suitable for various industrial and commercial uses.
  • Rare earth magnet material.
  • Countersunk design.
  • 0.750 in diameter.
  • 0.125 in height.
  • Maximum pull force of 8.6 lb.
  • Maximum operating temperature of 180 °F.
  • Nickel plated finish.
  • N42 grade.
Catalog No. 50-005-15769 Supplier MSC Supplier No. 95769360
This Neodymium disc magnet is designed for strong holding applications. It features a countersunk design for flush mounting and offers a high pull force, making it suitable for various industrial and commercial uses.
  • Rare earth magnet material.
  • Countersunk design.
  • 0.750 in diameter.
  • 0.125 in height.
  • Maximum pull force of 8.6 lb.
  • Maximum operating temperature of 180 °F.
  • Nickel plated finish.
  • N42 grade.