Learn More
MSC Mag-Mate Pad Eye, 17/32 in ID, 3,000 Lb Capacity, Carbon Steel
Catalog No. 5000528407
Encompass
This pad eye features a 17/32 inch inner diameter and a 3,000 pound capacity, constructed from durable carbon steel.
  • 17/32 inch inner diameter
  • 3,000 pound work load limit
  • Constructed from carbon steel
  • Overall length of 70 inches
  • Overall thickness of 16 mil
  • Overall height of 40 mm
Catalog No. 50-005-28407 Supplier MSC Supplier No. 30528640
This pad eye features a 17/32 inch inner diameter and a 3,000 pound capacity, constructed from durable carbon steel.
  • 17/32 inch inner diameter
  • 3,000 pound work load limit
  • Constructed from carbon steel
  • Overall length of 70 inches
  • Overall thickness of 16 mil
  • Overall height of 40 mm