Learn More
MSC Mag-Mate IMS123 36" Overall Length, Round Stainless Steel Inspection Mirror 2-1/4" Diam, 33-3/4" Handle, 7" Collapsed Length, Telescoping
Catalog No. NC1698669
Encompass
Mag-Mate IMS123 36" Overall Length, Round Stainless Steel Inspection Mirror 2-1/4" Diam, 33-3/4" Handle, 7" Collapsed Length, Telescoping
Catalog No. NC1698669 Supplier MSC Supplier No. 66307638
Mag-Mate IMS123 36" Overall Length, Round Stainless Steel Inspection Mirror 2-1/4" Diam, 33-3/4" Handle, 7" Collapsed Length, Telescoping