Learn More
MSC MAG-MATE Flexible Magnet Material Roll With Outdoor Adhesive 0.030"th x 1"wd x 25' lg
Supplier:  MSC 36576189
Encompass
300" Long x 1" Wide x 1/32" Thick Flexible Magnetic Strip 8 Lb Max Pull Force, 4 Linear Ft/Lb Magnetic Pull Force, Adhesive Back, Dark Brown
Catalog No. 50-190-9686