Learn More
V&P Scientific Inc MAGNETIC RACK, 1 Centrifuge Tube, 15mL or 50mL Volume
Catalog No. NC0457051
Encompass
MAGNETIC BEAD SEPARATION RACK, 1 Centrifuge Tube, 15mL or 50mL Volume, 1 Sided Bead Pellet Location, Low Position, 48MGO, NdFeB Magnet, 29.2mm Hole Diameter, HDPE (High Density Polyethylene)
Catalog No. NC0457051 Supplier V&P Scientific Inc Supplier No. VP 772FB1
MAGNETIC BEAD SEPARATION RACK, 1 Centrifuge Tube, 15mL or 50mL Volume, 1 Sided Bead Pellet Location, Low Position, 48MGO, NdFeB Magnet, 29.2mm Hole Diameter, HDPE (High Density Polyethylene)