Learn More
V&P Scientific Inc MAGNETIC PLATE, 96 Well, Standard or Deep Well, Round or Conical Bottom, 24 Round Posts, No Registration Base
Catalog No. NC1887378
Encompass
MAGNETIC BEAD SEPARATION PLATE, 96 Well, Standard or Deep Well Plate, Round or Conical Bottom, 24 Round Posts, 52MGO, NdFeB Magnets, Red Anodized Aluminum Magnet Frame, 1 Sided Pellet Location, 6.4mm Post Diameter, Axial - North Poles Up Magnetic Orientation, Gripper Accessible, SLAS Footprint, No Registration Base
Catalog No. NC1887378 Supplier V&P Scientific Inc Supplier No. VP 771LA1ZS
MAGNETIC BEAD SEPARATION PLATE, 96 Well, Standard or Deep Well Plate, Round or Conical Bottom, 24 Round Posts, 52MGO, NdFeB Magnets, Red Anodized Aluminum Magnet Frame, 1 Sided Pellet Location, 6.4mm Post Diameter, Axial - North Poles Up Magnetic Orientation, Gripper Accessible, SLAS Footprint, No Registration Base