Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC MAG/Siglec-4a protein, mouse | 100UG
SDP

Catalog No. 5000174621
Encompass_Preferred

Recombinant mouse myelin-associated glycoprotein (MAG/Siglec-4a) produced in HEK293 cells and supplied as a lyophilized, C-terminal 6xHis-tagged protein for biochemical and cell-based assays. Formulated from HEPES or phosphate buffer, the product has a reported molecular weight of 70-120 kDa and demonstrated activity in SH-SY5Y inhibition assays. Store frozen and aliquot to prevent repeated freeze-thaw.

  • Recombinant mouse MAG/Siglec-4a for functional assays.
  • Expressed in HEK293 to preserve mammalian post-translational modifications.
  • C-terminal 6xHis tag for purification and detection.
  • Lyophilized from buffered saline for stability and transport.
  • Reported molecular weight 70-120 kDa.
  • Demonstrated biological activity (ED50 ~3.13 μg/mL in SH-SY5Y assays).
  • Store at -20°C and aliquot to minimize freeze-thaw cycles.

Catalog No. 50-001-74621 Supplier Medchemexpress LLC Supplier No. HYP70319100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant mouse myelin-associated glycoprotein (MAG/Siglec-4a) produced in HEK293 cells and supplied as a lyophilized, C-terminal 6xHis-tagged protein for biochemical and cell-based assays. Formulated from HEPES or phosphate buffer, the product has a reported molecular weight of 70-120 kDa and demonstrated activity in SH-SY5Y inhibition assays. Store frozen and aliquot to prevent repeated freeze-thaw.

  • Recombinant mouse MAG/Siglec-4a for functional assays.
  • Expressed in HEK293 to preserve mammalian post-translational modifications.
  • C-terminal 6xHis tag for purification and detection.
  • Lyophilized from buffered saline for stability and transport.
  • Reported molecular weight 70-120 kDa.
  • Demonstrated biological activity (ED50 ~3.13 μg/mL in SH-SY5Y assays).
  • Store at -20°C and aliquot to minimize freeze-thaw cycles.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.