Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC MAG/Siglec-4a protein, rat (HEK293, His) | 100UG
SDP

Catalog No. 5000175075
Encompass_Preferred

Recombinant rat MAG/Siglec-4a protein expressed in HEK293 cells and purified with a C-terminal His tag; supplied as a lyophilized, glycosylated protein (~75-95 kDa) formulated in PBS with 8% trehalose. The product has low endotoxin (<0.1 EU/μg), shows biological activity in cell-based assays (ED50 2.501 μg/mL), and is intended for in vitro biochemical and cell-based studies. Recommended reconstitution is ≥100 μg/mL; store frozen for long-term stability.

  • Expression system: HEK293 mammalian cells
  • Tag: C-terminal His tag
  • Formulation: lyophilized from PBS with 8% trehalose
  • Endotoxin level: <0.1 EU/μg
  • Recommended reconstitution: not less than 100 μg/mL in PBS
  • Storage: store at -20°C or -80°C in aliquots for extended storage
  • Appearance: lyophilized powder

Catalog No. 50-001-75075 Supplier Medchemexpress LLC Supplier No. HYP702522100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Recombinant rat MAG/Siglec-4a protein expressed in HEK293 cells and purified with a C-terminal His tag; supplied as a lyophilized, glycosylated protein (~75-95 kDa) formulated in PBS with 8% trehalose. The product has low endotoxin (<0.1 EU/μg), shows biological activity in cell-based assays (ED50 2.501 μg/mL), and is intended for in vitro biochemical and cell-based studies. Recommended reconstitution is ≥100 μg/mL; store frozen for long-term stability.

  • Expression system: HEK293 mammalian cells
  • Tag: C-terminal His tag
  • Formulation: lyophilized from PBS with 8% trehalose
  • Endotoxin level: <0.1 EU/μg
  • Recommended reconstitution: not less than 100 μg/mL in PBS
  • Storage: store at -20°C or -80°C in aliquots for extended storage
  • Appearance: lyophilized powder

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.