Learn More
Research Products International Corp Mag Stir Genie Magnetic Stirrer 8 x 6 1/2 x 2
SDP
Catalog No. 501977043
Encompass
These innovative stirrers allow custom applications ranging from gentle stirring for cell cultures to aggressive mixing of viscous liquids. Four memory profiles can be stored that control speed, ramp rate, stir direction, time and powder level. Automatic mode allows recall of a stored program. A manual mode can also be selected that provides stir, stir reverse and time functions. The multi-position model features synchronized operation of all four stirring surfaces. This ensures the exact mixing parameters for each vessel and reduces error. These stirrers can be used in a cold room at 0°C or in an incubator up to 38°C.
Two year warranty.
Catalog No. 50-197-7043 Supplier Research Products International Corp Supplier No. 150600
These innovative stirrers allow custom applications ranging from gentle stirring for cell cultures to aggressive mixing of viscous liquids. Four memory profiles can be stored that control speed, ramp rate, stir direction, time and powder level. Automatic mode allows recall of a stored program. A manual mode can also be selected that provides stir, stir reverse and time functions. The multi-position model features synchronized operation of all four stirring surfaces. This ensures the exact mixing parameters for each vessel and reduces error. These stirrers can be used in a cold room at 0°C or in an incubator up to 38°C.
Two year warranty.