Learn More
Grainger MAG TPE 1 IN WX50 FT L MASTER
Catalog No. NC9302536
Encompass_Preferred
Magnetic Tape, 1 in. W x 50 ft. L, adhesive on one side, A magnetic surface on the other, Resistant to water and most detergents, Temperature range 70 deg.F or higher when applying, 0 to 150 deg.F afterward, 1/RL
This item is not returnable. View return policy
Catalog No. NC9302536 Supplier Grainger Supplier No. 3NJX3
Magnetic Tape, 1 in. W x 50 ft. L, adhesive on one side, A magnetic surface on the other, Resistant to water and most detergents, Temperature range 70 deg.F or higher when applying, 0 to 150 deg.F afterward, 1/RL