Learn More
Instech Vascular Access Button™ for Rat, Magnetic, One Channel 27ga (white port) for Intrathecal Catheters, with Injector
SDP
Catalog No. NC2816933
Encompass
This transcutaneous button features an external port with a septum for quick, clean and repeated access to a rat's implanted catheter. Applications include bolus or continuous dosing, blood sampling and IV self-administration. For intermittent access use a syringe fitted with a needlefree PinPort™ injector; for continuous access from outside the cage snap in a mating tether.
Group house animals by using the protective aluminum cap — an animal welfare benefit that has never before been available to rats with externalized catheters.
Catalog No. NC2816933 Supplier Instech Supplier No. VABR1B/27
This transcutaneous button features an external port with a septum for quick, clean and repeated access to a rat's implanted catheter. Applications include bolus or continuous dosing, blood sampling and IV self-administration. For intermittent access use a syringe fitted with a needlefree PinPort™ injector; for continuous access from outside the cage snap in a mating tether.
Group house animals by using the protective aluminum cap — an animal welfare benefit that has never before been available to rats with externalized catheters.