Learn More
Medchemexpress LLC Magainin 1 | 108433-99-4 | 99.9% | 2409.85 | 500 UG
SDP
Catalog No. 5000424834
Encompass_Preferred
Magainin 1 is an antimicrobial and amphipathic peptide isolated from the skin of *Xenopus laevis*. It exhibits antibiotic activity against various Gram-negative and Gram-positive bacteria by interacting with acidic lipids to form an amphiphilic helix, leading to membrane leakage.
  • Antimicrobial and amphipathic peptide
  • Isolated from *Xenopus laevis* skin
  • Exhibits antibiotic activity against Gram-negative and Gram-positive bacteria
  • Interacts with acidic lipids through electrostatic and hydrophobic interactions
  • Forms an amphiphilic helix
  • Induces leakage of calcein from negatively-charged vesicles
  • Binds to bovine brain phosphatidylserine sonicated vesicles
Catalog No. 50-004-24834 Supplier Medchemexpress LLC Supplier No. HYP0269500UG
Magainin 1 is an antimicrobial and amphipathic peptide isolated from the skin of *Xenopus laevis*. It exhibits antibiotic activity against various Gram-negative and Gram-positive bacteria by interacting with acidic lipids to form an amphiphilic helix, leading to membrane leakage.
  • Antimicrobial and amphipathic peptide
  • Isolated from *Xenopus laevis* skin
  • Exhibits antibiotic activity against Gram-negative and Gram-positive bacteria
  • Interacts with acidic lipids through electrostatic and hydrophobic interactions
  • Forms an amphiphilic helix
  • Induces leakage of calcein from negatively-charged vesicles
  • Binds to bovine brain phosphatidylserine sonicated vesicles