Learn More
Cayman Chemical Magainin 1 (trifluoroacetate salt), Purity: ≥95%, 10 mg
SDP
Catalog No. 502910286
Encompass_Preferred
An antimicrobial cationic peptide; reduces germination percentage and mean tube elongation in kiwi fruit pollen (EC50s = 14 and 15.8 µM, respectively); decreases viral replication in Vero cells infected with HSV-1 or HSV-2 (EC50s = 36.59 and 33.2 µM, respectively) without affecting cell viability (CC50 = >100 µM); induces apoptosis in HL-60 cells at 10 and 50 µM and hemolysis in isolated human erythrocytes at 50 µM; induces DNA fragmentation in HL-60 cells in a reporter assay at 10 and 50 µM
Catalog No. 50-291-0286 Supplier Cayman Chemical Supplier No. 3565110 MG
An antimicrobial cationic peptide; reduces germination percentage and mean tube elongation in kiwi fruit pollen (EC50s = 14 and 15.8 µM, respectively); decreases viral replication in Vero cells infected with HSV-1 or HSV-2 (EC50s = 36.59 and 33.2 µM, respectively) without affecting cell viability (CC50 = >100 µM); induces apoptosis in HL-60 cells at 10 and 50 µM and hemolysis in isolated human erythrocytes at 50 µM; induces DNA fragmentation in HL-60 cells in a reporter assay at 10 and 50 µM