Learn More
CPC Scientific H-Gly-Ile-Gly-Lys-Phe-Leu-His-Ser-Ala-Lys-Lys-Phe-Gly-Lys-Ala-Phe-Val-Gly-Glu-Ile-Met-Asn-Ser-OH (trifluoroacetate salt) 0.5MG
SDP
Catalog No. 502108820
Encompass
A hemolytic and antimicrobial peptide isolated from frog skin. This naturally occurring peptide causes a change in membrane permeability and forms cation-selective channels at concentrations over 2 µM.ONE-LETTER SEQUENCE: GIGKFLHSAKKFGKAFVGEIMNSMOLECULAR FORMULA: C114H180N30O29S1MOLECULAR WEIGHT:2466.93STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [108433-95-0], SYNONYMS: Magainin IIRESEARCH AREA: AntimicrobialREFERENCES: A. Mor et al., Biochemistry, 30, 8824 (1991)
Catalog No. 50-210-8820 Supplier CPC Scientific Supplier No. ATMP007A
A hemolytic and antimicrobial peptide isolated from frog skin. This naturally occurring peptide causes a change in membrane permeability and forms cation-selective channels at concentrations over 2 µM.ONE-LETTER SEQUENCE: GIGKFLHSAKKFGKAFVGEIMNSMOLECULAR FORMULA: C114H180N30O29S1MOLECULAR WEIGHT:2466.93STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [108433-95-0], SYNONYMS: Magainin IIRESEARCH AREA: AntimicrobialREFERENCES: A. Mor et al., Biochemistry, 30, 8824 (1991)