Learn More
Medchemexpress LLC Magainin 2 | 108433-95-0 | 98.8% | 2466.90 | 500 UG
SDP
Catalog No. 5000424904
Encompass_Preferred
Magainin 2, an antimicrobial peptide isolated from the skin of the African clawed frog, Xenopus laevis, exhibits antibiotic activity against numerous gram-negative and gram-positive bacteria, as well as protozoa. It exerts cytotoxicity by interacting with anionic phospholipids abundant in bacterial membranes, permeabilizing cell membranes with low toxicity to mammalian cells.
  • Exhibits broad-spectrum antibiotic activity against bacteria and protozoa
  • Interacts with anionic phospholipids in bacterial membranes
  • Induces morphological changes in Escherichia coli
  • Permeabilizes bacterial cell membranes without significant toxicity to mammalian cells
  • Disrupts osmotic balance in protozoa like Paramecium caudatum
Catalog No. 50-004-24904 Supplier Medchemexpress LLC Supplier No. HYP0270500UG
Magainin 2, an antimicrobial peptide isolated from the skin of the African clawed frog, Xenopus laevis, exhibits antibiotic activity against numerous gram-negative and gram-positive bacteria, as well as protozoa. It exerts cytotoxicity by interacting with anionic phospholipids abundant in bacterial membranes, permeabilizing cell membranes with low toxicity to mammalian cells.
  • Exhibits broad-spectrum antibiotic activity against bacteria and protozoa
  • Interacts with anionic phospholipids in bacterial membranes
  • Induces morphological changes in Escherichia coli
  • Permeabilizes bacterial cell membranes without significant toxicity to mammalian cells
  • Disrupts osmotic balance in protozoa like Paramecium caudatum