Learn More
Cayman Chemical MagaInIn 2trIfluoroacetAT 10mg
SDP
Catalog No. 502914854
Encompass_Preferred
An antimicrobial cationic peptide; active against the bacteria E. coli, K. pneumoniae, S. epidermidis, S. aureus, and the fungus C. albicans (MICs = 5, 10, 10, 50, and 80 UG/ml, respectively); reduces germination percentage and mean tube elongation in kiwi fruit pollen at 20 µM; decreases viral replication in Vero cells infected with herpes simplex virus 1 (HSV-1) or HSV-2 (EC50s = 22.16 and 19.8 µM, respectively) without affecting cell viability (CC50 = >100 µM)
Catalog No. 50-291-4854 Supplier Cayman Chemical Supplier No. 3690010 MG
An antimicrobial cationic peptide; active against the bacteria E. coli, K. pneumoniae, S. epidermidis, S. aureus, and the fungus C. albicans (MICs = 5, 10, 10, 50, and 80 UG/ml, respectively); reduces germination percentage and mean tube elongation in kiwi fruit pollen at 20 µM; decreases viral replication in Vero cells infected with herpes simplex virus 1 (HSV-1) or HSV-2 (EC50s = 22.16 and 19.8 µM, respectively) without affecting cell viability (CC50 = >100 µM)