Learn More
CPC Scientific H-Gly-Ile-Gly-Lys-Phe-Leu-His-Ser-Ala-Gly-Lys-Phe-Gly-Lys-Ala-Phe-Val-Gly-Glu-Ile-Met-Lys-Ser-OH (trifluoroacetate salt) 1MG
SDP
Catalog No. 502108823
Encompass
This peptide originally isolated from frog skin, with antimicrobial activity.ONE-LETTER SEQUENCE: GIGKFLHSAGKFGKAFVGEIMKSMOLECULAR FORMULA: C112H177N29O28S1MOLECULAR WEIGHT:2409.88STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [108433-99-4], RESEARCH AREA: AntimicrobialREFERENCES: M.Zasloff, Proc. Natl. Acad. Sci. USA, 84, 5449 (1987);
Catalog No. 50-210-8823 Supplier CPC Scientific Supplier No. ATMP008B
This peptide originally isolated from frog skin, with antimicrobial activity.ONE-LETTER SEQUENCE: GIGKFLHSAGKFGKAFVGEIMKSMOLECULAR FORMULA: C112H177N29O28S1MOLECULAR WEIGHT:2409.88STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [108433-99-4], RESEARCH AREA: AntimicrobialREFERENCES: M.Zasloff, Proc. Natl. Acad. Sci. USA, 84, 5449 (1987);