Learn More
CPC Scientific H-Asp-Ala-Glu-Ala-Val-Gly-Pro-Glu-Ala-Phe-Ala-Asp-Gln-Asp-Leu-Asp-Glu-Arg-Glu-Val-Arg-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific ATBP005B
Encompass
ONE-LETTER SEQUENCE: DAEAVGPEAFADQDLDEREVRMOLECULAR FORMULA: C97H150N28O39MOLECULAR WEIGHT:2332.4STORAGE CONDITIONS: -20 5CRESEARCH AREA: Antimicrobial
Catalog No. 50-210-8800