Learn More
Perkin Elmer US LLC Magazine for Liquid Sample Sealer
Catalog No. 502112713
Encompass_Preferred
The Liquid Sample Sealer is used to seal volatile, non-viscous, non-corrosive samples in either the 4 µL aluminum capsules (02400642) or 30 µL aluminum capsules (00090709).
Catalog No. 50-211-2713 Supplier Perkin Elmer US LLC Supplier No. B0501078
The Liquid Sample Sealer is used to seal volatile, non-viscous, non-corrosive samples in either the 4 µL aluminum capsules (02400642) or 30 µL aluminum capsules (00090709).