Learn More
V&P Scientific Inc MAGNETIC PLATE, 96 Well, Standard, Flat Bottom, 96 Round Posts, BioTek Plate Washer Compatible
Catalog No. NC2309030
Encompass
MAGNETIC BEAD SEPARATION PLATE, 96 Well, Standard Plate, Flat Bottom, 96 Round Posts, 52MGO, NdFeB Magnets, Red Anodized Aluminum Magnet Frame, Bottom Center Pellet Location, 6.4mm Post Diameter, Axial - North Poles Up Magnetic Orientation, Gripper Accessible, BioTek Plate Washer Compatible, Non-SLAS Footprint, No Registration Base
Catalog No. NC2309030 Supplier V&P Scientific Inc Supplier No. VP 771BTR2Z
MAGNETIC BEAD SEPARATION PLATE, 96 Well, Standard Plate, Flat Bottom, 96 Round Posts, 52MGO, NdFeB Magnets, Red Anodized Aluminum Magnet Frame, Bottom Center Pellet Location, 6.4mm Post Diameter, Axial - North Poles Up Magnetic Orientation, Gripper Accessible, BioTek Plate Washer Compatible, Non-SLAS Footprint, No Registration Base