Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals MagBeads FastDNA Kit

Catalog No. MP119612096 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
No. of Reactions:
96 Preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
MP119612096 96 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP119612096 Supplier MP Biomedicals Supplier No. 119612096
Add to Cart
Add to Cart

Produces pure, high-quality genomic DNA suitable for downstream molecular biology applications including PCR and qPCR.

  • MagBeads FastDNA Kit (Ready-to-Use for MagFlex-96) is designed for quick and convenient extraction of genomic DNA from various samples such as animal tissue, cells, blood, saliva, swabs, blood spots, and semen, with the use of magnetic beads nucleic acid purification technology
  • The extracted DNA is of high quality and purity, suitable for various downstream molecular biology applications such as PCR and qPCR
TRUSTED_SUSTAINABILITY

Specifications

Format Kit
Isolation Technology Magnetic Bead
Scale Mini
No. of Reactions 96 Preps
Product Type MagBeads FastDNA Kit
Voltage 0/1 V
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.