Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals MagBeads FastDNA Kit for Blood

Catalog No. MP119610096 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
No. of Reactions:
96 Preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
MP119610096 96 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP119610096 Supplier MP Biomedicals Supplier No. 119610096
Add to Cart
Add to Cart

Compatible with the MagFlex-96 aNAP System, enabling simultaneous processing of up to 96 samples.

  • The MagBeads FastDNA Kit for Blood offers an efficient solution for genomic DNA purification from various blood samples, including fresh and frozen human whole blood preserved in common anticoagulants, buffy coat, and saliva
  • Designed to be compatible with the Magflex-96 System, it supports high-throughput processing of up to 96 samples and accommodates varying sample volumes from 50 μL to 500 μL
  • The kit features a rapid, integrated approach for DNA isolation by combining lysis and binding steps, omitting the need for additional sample processing steps
  • The Magnetic Beads used in MagBeads FastDNA Kit for Blood simplifies the DNA purification process by eliminating complicated RNA removal step, ensuring highly efficient and specific binding of DNA
  • Specialized wash buffers included in the kit deliver high washing efficiency, removing proteins, salts, and other impurities through a quick, streamlined wash procedure
  • The DNA eluted in ionic strength elution buffer is of high quality, free from contaminants, and fully compatible with downstream applications such as PCR and other molecular techniques
  • Integrated lysis and binding steps, combined with simplified magnetic bead technology, eliminate the need for RNA removal and reduce processing time to just 60 minutes
  • Delivers up to 50 μg DNA yield with elution volumes of 100-200 μL, ensuring concentrated, high-quality DNA for downstream applications.
TRUSTED_SUSTAINABILITY

Specifications

Format Kit
Isolation Technology Magnetic Bead
Scale Mini
No. of Reactions 96 Preps
Product Type MagBeads FastDNA Kit
Voltage 0/1 V
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.