Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals MagBeads FastDNA Kit for Feces

Catalog No. MP119608096 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
No. of Reactions:
96 Preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
MP119608096 96 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP119608096 Supplier MP Biomedicals Supplier No. 119608096
Add to Cart
Add to Cart

Processes both fresh and frozen human and animal feces with complete DNA extraction in under 60 minutes.

  • The MagBeads FastDNA Kit for Feces allows quick and efficient isolation of high-quality genomic DNA from fresh or frozen human and animal feces in less than 60 mins
  • Samples are placed into Lysing Matrix E tube and used with FastPrep™ Instruments from MP Biomedicals to effectively lyse host cells as well as bacteria, fungi, viruses, protists and other cells present in fecal samples within 40 seconds
  • The kit is also compatible with most of the automated nucleic acid extraction instruments on the market or it can be operated manually
  • It consists of specially formulated reagents to eliminate humic acid, polysaccharides, phenolic compounds, and enzyme inhibitors from feces and thus allowing extraction of highly pure genomic DNA ready for PCR, restriction digestion, electrophoresis, and other desired applications
TRUSTED_SUSTAINABILITY

Specifications

Format Kit
Isolation Technology Magnetic Bead
Scale Mini
No. of Reactions 96 Preps
Product Type MagBeads FastDNA Kit
Voltage 0/1 V
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.