Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ MagBeads FastDNA™ Kit for Feces

Catalog No. MP116570400 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
MagBeads FastDNA™ Kit for Feces
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
MP116570400 MagBeads FastDNA™ Kit for Feces
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP116570400 Supplier MP Biomedicals™ Supplier No. 116570400
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The kit allows for the isolation of genomic DNA. Samples are placed into Lysing Matrix E tubes and used with FastPrep™ instruments to lyse host cells, bacteria, fungi, viruses, protists and other cells.

  • Quick and efficient isolation of high-quality genomic DNA from:
    • Fresh or frozen human and animal feces, intestinal contents
      • < 60 min. for extraction
      • High yield and throughput
      • Free from inhibitors
  • Samples placed into Lysing Matrix E tubes and used with FastPrep™ instruments to lyse within 40 seconds:
    • Host cells
    • Bacteria
    • Fungi
    • Viruses
    • Protists
    • Other cells in fecal samples

Kit:

  • Compatible with most automated nucleic acid extraction instruments
  • Can be operated manually
  • No phenol/chloroform or other toxic chemicals
  • Consists of specially formulated reagents to eliminate:
    • Humic acid
    • Polysaccharides
    • Phenolic compounds
    • Enzyme inhibitors from feces
  • Allows for extraction of highly pure genomic DNA ready for:
    • PCR
    • Restriction digestion
    • Electrophoresis
TRUSTED_SUSTAINABILITY

Specifications

Detection Method Magnetic Extraction
Format Kit
Isolation Technology Magnetic Particles
Sample Type Feces
For Use With (Equipment) Homogenizer such as FastPrep™-24 5G for automated extractions
For Use With (Application) DNA Extraction
No. of Reactions 50 Preps
Product Type DNA Extraction Kit
Target DNA
Test Time <60 min.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.