Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals MagBeads FastDNA™ Kit for FFPE

Catalog No. MP116576096 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Scale:
Mini
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Scale
MP116576096 Mini
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP116576096 Supplier MP Biomedicals Supplier No. 116576096
Add to Cart
Add to Cart

MagBeads FastDNA™ Kit for FFPE is intended for rapid extraction of DNA from FFPE sample.

  • MagBeads FastDNA® Kit for FFPE is based on the purification method of high binding magnetic particles. The sample is lysed and digested. DNA is released into the lysate.
  • After addition of magnetic particles and binding solution, DNA will be adsorbed on the surface of magnetic particles, and impurities such as proteins will be removed without adsorption.
  • The adsorbed particles were washed with washing buffer to remove the proteins and impurities, washed with ethanol to remove salts, and finally the DNA was eluted with Elution Buffer.
  • This kit uses two combination methods. High-salt Bind is conducive to remove pigments or polysaccharides from complex FFPE samples, so as to improve the purity of nucleic acid and avoid blocking aligent 2100.
  • Alcohol mediated adsorption is conducive for improving the nucleic acid yield.
TRUSTED_SUSTAINABILITY

Specifications

Format Kit
Isolation Technology Magnetic Bead
Scale Mini
Sample Type FFPE
Product Type MagBeads FastDNA™ Kit
Voltage 0/1 V
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.