Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals MagBeads FastDNA Kit for Microbiome

Catalog No. MP119613096 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
No. of Reactions:
96 Preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
MP119613096 96 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP119613096 Supplier MP Biomedicals Supplier No. 119613096
Add to Cart
Add to Cart

Delivers high-quality genomic DNA suitable for PCR, qRT-PCR, and metagenomic library construction for comprehensive genomic analyses.

  • The MagBeads FastDNA Kit for Microbiome is an advanced genomic DNA (gDNA) extraction solution based on the next-generation silicon-based magnetic bead technology
  • This kit is designed to deliver high-fidelity gDNA extraction from a diverse array of biological specimens, encompassing bacterial communities, a spectrum of bodily fluids, bronchoalveolar lavage fluids, and environmental samples such as soil
  • The kit integrates an effective lysis mechanism, ensuring thorough cell lysis and subsequent DNA purification across a broad spectrum of microorganisms, including bacteria, archaea, and fungi
  • The resulting gDNA is of high purity and suitable for numerous downstream molecular applications, such as Polymerase Chain Reaction (PCR), Quantitative Real-Time PCR (qRT-PCR), and metagenomic library construction, facilitating comprehensive genomic analyses
TRUSTED_SUSTAINABILITY

Specifications

Format Kit
Isolation Technology Magnetic Bead
Scale Mini
No. of Reactions 96 Preps
Product Type MagBeads FastDNA Kit
Voltage 0/1 V
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.