Learn More
Description
- The MagBeads FastDNA™ Kit for Soil allows quick and efficient isolation of high-quality genomic DNA from soil in less than 60 mins
- Samples are placed into Lysing Matrix E tubes and used with FastPrep™ Instruments from MP Biomedicals to effectively lyse within 40 seconds:
- Host cells
- Bacteria
- Fungi
- Viruses
- Protists
- Other cells present in soil samples
- The kit is also compatible with most of the automated nucleic acid extraction instruments on the market or it can be operated manually. It consists of specially formulated reagents to eliminate:
- Humic acid
- Polysaccharides
- Phenolic compounds
- Enzyme inhibitors from soil
- It allows for extraction of highly pure genomic DNA ready for:
- PCR
- Restriction digestion
- Electrophoresis
- Other desired applications
Specifications
Specifications
| Format | Kit |
| Isolation Technology | Magnetic Bead |
| Scale | Mini |
| Sample Type | Soil (Various Types) |
| Sequencing Type | DNA and RNA Sequencing |
| Product Type | DNA Isolation Kit |
| Quantity | 50 Preps |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.