Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals MagBeads FastDNA/RNA Kit for Pathogen

Catalog No. MP116589050 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Isolation Technology:
Magnetic Bead
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Isolation Technology
MP116589050 Magnetic Bead
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP116589050 Supplier MP Biomedicals Supplier No. 116589050
Add to Cart
Add to Cart

Single liquid-transfer step reduces hands-on time, minimizes human error, and boosts sample throughput. Effectively prevents exogenous nucleic acid contamination, guaranteeing accurate and reliable sample composition.

  • The MagBeads FastDNA/RNA Kit for Pathogen offers an advanced rapid nucleic acid extraction solution, based on the next-generation silicon-based magnetic bead technology.
  • This kit is designed to deliver high-fidelity gDNA/RNA extraction from samples, such as bodily fluids, for detection requirements.
  • The kit features a streamlined extraction protocol that combines rapid processing with operational reliability, requiring only a single liquid-transfer step prior to automation.
  • Complementing this efficiency, the integrated optimized lysis protocol ensures complete and consistent bacterial cell disruption, establishing a robust foundation for highly reliable nucleic acid detection outcomes.
  • To guarantee accurate results, this product effectively prevents exogenous nucleic acid contamination, thereby providing an accurate reflection of the sample's intrinsic species composition.
  • The nucleic acid isolated through this methodology is characterized by its high purity, making it suitable for a wide array of downstream molecular applications.
  • It is particularly well-suited for applications such as PCR, RT-PCR, and the construction of metagenomic libraries.
  • Available in prefilled formats - Ready-to-use for MPure-32™ (Cat. No. 117040500) and Ready-to-use for MPure-96™ (Cat. No. 117040600) aNAP Systems.
TRUSTED_SUSTAINABILITY

Specifications

Format Kit
Isolation Technology Magnetic Bead
Scale Mini
Sample Type Pathogens
Product Type MagBeads FastDNA/RNA Kit
Voltage 0/1 V
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.