Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals MagBeads FastRNA Kit for Blood

Catalog No. MP119609096 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
No. of Reactions:
96 Preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
MP119609096 96 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP119609096 Supplier MP Biomedicals Supplier No. 119609096
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Scalable, automatable magnetic bead-based RNA purification compatible with MagFlex-96.

  • MagBeads FastRNA Kit for Blood (Ready-To-Use for MagFlex-96) is a scalable and automatable magnetic bead-based RNA purification technology that is designed for total isolation of RNA from whole blood preserved in different anticoagulants (EDTA, Heparin and sodium citrate)
  • RNA degradation prevention is the backbone of RNA purification
  • The use of our specially formulated Buffer MBL containing huge amounts of chaotropic ions omits the need of excessive homogenization and enables highly efficient lysis with the immediate inactivation of RNases, which are typically present in most biological materials
  • The MagBeads FastRNA Kit for Blood simplifies the RNA purification process by eliminating the need for complicated erythrocyte lysis and isolation of leukocyte pellet, making it hassle-free and time efficient
  • Contaminated DNA is removed by a DNase I treatment step
  • Simplified dual washing steps largely remove salts, metabolites, and macromolecular cellular components
  • Pure RNA extracts of high yield and purity, compatible with downstream applications such as RT-qPCR amplification, are then eluted under low ionic strength conditions with RNase-free water
TRUSTED_SUSTAINABILITY

Specifications

Buffer Lysis Buffer, Binding Buffer, Wash Buffer 1 and 2, Wash Buffer 1 and 2, Elution Buffer
Isolation Technology Magnetic Bead
Species All
Sample Type MagBeads FastRNA Kit
No. of Reactions 96 Preps
Product Type MagBeads FastRNA Kit
Sterility Non-sterile
Target Total RNA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.