Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals MagBeads FastRNA Kit for Feces

Catalog No. MP116588050 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Target:
Total RNA
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Target
MP116588050 Total RNA
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP116588050 Supplier MP Biomedicals Supplier No. 116588050
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Yields up to 250 μg of total RNA from up to 200 mg of fecal samples. Simplifies RNA purification, saving time and minimizing handling steps.

  • The MagBeads FastRNA Kit for Feces is a scalable and automatable magnetic bead-based RNA purification technology, designed for the efficient isolation of total RNA from fecal samples.
  • Available in prefilled formats – Ready-to-use for MPure-32™ (Cat. No. 117040300) and Ready-to-use for MPure-96™ (Cat. No. 117040400) aNAP Systems.
  • The composition of fecal samples is largely dependent on diet, which includes fibers, undigested particles, bilirubin, complex polysaccharides, and lipids. These compounds impair sample homogenization, decreasing both the quantity and quality of the extracted RNA. The MagBeads FastRNA Kit for Feces integrates our
  • state-of-the-art beads-beating technology, a lysis chemistry compatible with the widest range of samples and our proprietary inhibitor removal expertise. A new technology of selective binding of RNA along with our FastDNase I Enzyme treatment allows the complete removal of DNA. The MagBeads FastRNA Kit for Feces streamlines RNA purification by overcoming the challenges associated with diverse fecal sample types, making the process both hassle-free and time efficient. A DNase I treatment step effectively removes contaminated DNA, ensuring high RNA yield and purity. The extracted RNA is immediately compatible with downstream applications such as RT-qPCR amplification, eliminating the need for additional inhibitor or DNA removal steps.
TRUSTED_SUSTAINABILITY

Specifications

Buffer Lysis Buffer
Control Type RNA Control
Isolation Technology Magnetic Bead
Species All
Sample Type Feces
Product Type MagBeads FastRNA Kit
Sterility Non-sterile
Target Total RNA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.