Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals MagBeads FastRNA Kit for FFPE

Catalog No. MP116573192 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Scale:
Mini
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Scale
MP116573192 Mini
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP116573192 Supplier MP Biomedicals Supplier No. 116573192
Add to Cart
Add to Cart

MagBeads FastRNA Kit for Formalin-Fixed, Paraffin-Embedded, FFPE, is intended for rapid extraction of RNA from tissue, cells, blood, and other clinical samples. RNA can be used directly for RT-PCR, quantitative RT-PCR and so on.

  • MagBeads FastRNA Kit for FFPE is based on the purification method of high binding magnetic particles. The sample is lysed and digested.
  • RNA is released into the lysate.
  • After addition of magnetic particles and binding solution, RNA will be adsorbed on the surface of magnetic particles, and impurities such as proteins will be removed without adsorption.
  • The adsorbed particles were washed with washing buffer to remove the proteins and impurities, washed with ethanol to remove salts, and finally the RNA was eluted with Elution Buffer.
TRUSTED_SUSTAINABILITY

Specifications

Format Kit
Isolation Technology Magnetic Bead
Scale Mini
Sample Type FFPE
Product Type MagBeads FastRNA Kit
Voltage 0/1 V
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.