Learn More
Apexbio Technology LLC MagBeads PCR Purification Kit 100tests
SDP
Catalog No. 5000097192
Encompass_Preferred
The MagBeads PCR Purification Kit utilizes magnetic bead-based technology to selectively bind and purify DNA fragments from PCR mixtures The magnetic beads facilitate efficient separation of nucleic acids from proteins primers nucleotides and other contaminants through reversible binding under specific buffer conditions This mechanism enables rapid and high-yield recovery of DNA suitable for downstream applications such as sequencing cloning and genotyping The kit provides a streamlined workflow improving reproducibility and scalability in molecular biology and genomic research workflows
Catalog No. 50-000-97192 Supplier Apexbio Technology LLC Supplier No. K1625100
The MagBeads PCR Purification Kit utilizes magnetic bead-based technology to selectively bind and purify DNA fragments from PCR mixtures The magnetic beads facilitate efficient separation of nucleic acids from proteins primers nucleotides and other contaminants through reversible binding under specific buffer conditions This mechanism enables rapid and high-yield recovery of DNA suitable for downstream applications such as sequencing cloning and genotyping The kit provides a streamlined workflow improving reproducibility and scalability in molecular biology and genomic research workflows