Learn More
FUJIFILM Biosciences Exosome Isolation Kit, PS affinity, Magnetic beads, 10 tests
Catalog No. NC2044062
Encompass_Preferred
The MagCapture Exosome Isolation Kit PS Ver.2 is an affinity-based magnetic bead kit for isolating intact extracellular vesicles (EVs) from cell culture supernatant, serum, and plasma. The kit captures EVs via phosphatidylserine (PS) affinity and enables elution of intact vesicles for downstream molecular assays and uptake experiments, offering higher yield and purity compared with ultracentrifugation.
  • Affinity capture using phosphatidylserine-binding magnetic beads.
  • Elution yields intact extracellular vesicles suitable for downstream analysis.
  • Compatible with culture supernatant, serum, and plasma samples.
  • Manual magnetic-bead workflow; no ultracentrifugation required.
  • Packaged for 10 tests with buffers and reaction tubes included.
Catalog No. NC2044062 Supplier FUJIFILM Biosciences Supplier No. 29084103
The MagCapture Exosome Isolation Kit PS Ver.2 is an affinity-based magnetic bead kit for isolating intact extracellular vesicles (EVs) from cell culture supernatant, serum, and plasma. The kit captures EVs via phosphatidylserine (PS) affinity and enables elution of intact vesicles for downstream molecular assays and uptake experiments, offering higher yield and purity compared with ultracentrifugation.
  • Affinity capture using phosphatidylserine-binding magnetic beads.
  • Elution yields intact extracellular vesicles suitable for downstream analysis.
  • Compatible with culture supernatant, serum, and plasma samples.
  • Manual magnetic-bead workflow; no ultracentrifugation required.
  • Packaged for 10 tests with buffers and reaction tubes included.