Learn More
R&D Systems™ MagCellect Streptavidin Ferrofluid

Specifications
Specifications
| Content And Storage | Store the unopened product at 2 - 8° C. Do not use past expiration date. |
| For Use With (Application) | Cell Isolation |
| Product Type | Streptavidin Ferrofluid |
| Quantity | 1 Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.