Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Supertek™ Magdeburg Pressure Disks
GREENER_CHOICESCI_ED

Catalog No. S139433 Shop All Supertek Scientific Products
Change view
Click to view available options
Grade:
6+
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S139433 6+
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S139433 Supplier Supertek™ Supplier No. PH12184
Add to Cart
Add to Cart

When using the Supertek™ Madeburg Pressure Disks, join the hemispheres, hook them up to a vacuum pump and evacuate the air inside the hemispheres. What happens?

  • Kit consists of...
  • 2 - Plastic 3.94 in. (dia.) hemispheres with wooden handles.
  • One of the hemispheres has a raised edge to hold a rubber seal.
  • The other hemisphere has a stopcock for attaching to vacuum tubing.
  • Join the hemispheres and hook them up to a vacuum pump.
  • Evacuate the air inside the hemispheres making it hard to pull them apart.
  • Open the stopcock to let air in and the hemispheres will separate easily.
  • Instruction sheet included.

Specifications

Grade 6+
Format Kit
Languages English
Subject Physical Science - Pressure and Volume
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.