Learn More
Medchemexpress LLC MAGE-A1-derived peptide acetate | 99.6% | 1270.51 | 5 MG
SDP
Catalog No. 5000423724
Encompass_Preferred
MAGE-A1-derived peptide acetate is a short peptide sequence derived from the MAGE-A1 protein. It functions as a tumor-specific antigen, capable of being recognized and activated by cytotoxic T lymphocytes (CTLs). This activation initiates an immune response against tumor cells that express MAGE-A1, ultimately leading to their lysis and death. This compound is valuable for research in tumor immunity.
  • Derived from MAGE-A1 protein.
  • Acts as a tumor-specific antigen.
  • Activates cytotoxic T lymphocytes (CTLs).
  • Generates an immune response against MAGE-A1 expressing tumor cells.
  • Induces tumor cell lysis and death.
  • Used in tumor immunity studies.
Catalog No. 50-004-23724 Supplier Medchemexpress LLC Supplier No. HYP10496A5MG
MAGE-A1-derived peptide acetate is a short peptide sequence derived from the MAGE-A1 protein. It functions as a tumor-specific antigen, capable of being recognized and activated by cytotoxic T lymphocytes (CTLs). This activation initiates an immune response against tumor cells that express MAGE-A1, ultimately leading to their lysis and death. This compound is valuable for research in tumor immunity.
  • Derived from MAGE-A1 protein.
  • Acts as a tumor-specific antigen.
  • Activates cytotoxic T lymphocytes (CTLs).
  • Generates an immune response against MAGE-A1 expressing tumor cells.
  • Induces tumor cell lysis and death.
  • Used in tumor immunity studies.