Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 4100MSM1P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: MA454. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1 kappa. Positive Control: Melanoma cell lines. Melanomas or testicular carcinomas. Cellular Staining: Cytoplasmic. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Human MAGE-A1 full length recombinant protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 42-46kDa.Chromosome: Xq28. Gene Symbol: MAGEA1. Gene ID: 4100. Ensemble: ENSG00000198681. Uniprot: NP_004979.
Catalog No. 50-188-4916