Learn More
Aviva Systems Biology Corp MAGEA1 Antibody - C-terminal region (ARP46116_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46116_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against MAGEA1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human MAGEA1. Alternate names: MAGE1, MGC9326, CT1.1ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-59233