Learn More
Aviva Systems Biology Corp MAGEA11 Antibody - middle region (ARP59877_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59877_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MAGEA11. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human MAGEA11. Alternate names: CT1.11, MAGE-11, MAGE11, MAGEA-11, MGC10511ApplicationWB. Species reactivity: Human, Zebrafish
Catalog No. 50-286-863