Learn More
Aviva Systems Biology Corp MAGEA5 Antibody - N-terminal region (ARP46295_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46295_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MAGEA5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human MAGEA5. Alternate names: MAGE5, MAGEA4, MGC129526, CT1.5ApplicationWB. Species reactivity: Human
Catalog No. 50-285-247