Learn More
Aviva Systems Biology Corp MAGEA8 Antibody - N-terminal region (ARP42233_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42233_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MAGEA8. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human MAGEA8. Alternate names: MAGE8, MGC2182, CT1.8ApplicationIHC, WB. Species reactivity: Human
Catalog No. 50-014-58155