Learn More
Aviva Systems Biology Corp MAGEA9 Antibody - middle region (ARP38609_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38609_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MAGEA9. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human MAGEA9. Alternate names: MAGE9, MGC8421, CT1.9, MAGEA9BApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Rabbit, Rat
Catalog No. 50-014-56726