Learn More
Aviva Systems Biology Corp MAGEA9 Antibody - middle region (ARP35768_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35768_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MAGEA9. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human MAGEA9. Alternate names: MAGE9, MGC8421, CT1.9, MAGEA9BApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Pig, Rabbit, Rat, Yeast
Catalog No. 50-014-55843