Learn More
Aviva Systems Biology Corp MAGEB3 Antibody - N-terminal region (ARP56338_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56338_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MAGEB3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human MAGEB3. Alternate names: CT3.5ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-61819