Learn More
Aviva Systems Biology Corp MAGEB4 Antibody - N-terminal region (ARP56340_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56340_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MAGEB4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human MAGEB4. Alternate names: MGC33144, CT3.6ApplicationWB. Species reactivity: Human
Catalog No. 50-014-62988