Learn More
Aviva Systems Biology Corp MAGED2 Antibody - N-terminal region (ARP59887_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59887_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MAGED2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human MAGED2. Alternate names: 11B6, BCG1, HCA10, JCL-1, MAGE-D2, MAGED, MGC8386ApplicationWB. Species reactivity: Dog, Horse, Human, Pig, Rabbit
Catalog No. 50-286-866